Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentyl bromide, 98%

Catalog No. AC111600010
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111600010 1 L Glass bottle
AC111600500 50 mL Glass bottle
AC111602500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC111600010 Supplier Thermo Scientific Chemicals Supplier No. 111600010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 137-43-9 | C5H9Br | 149.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-43-9
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.03
MDL Number MFCD00001359
InChI Key BRTFVKHPEHKBQF-UHFFFAOYSA-N
Synonym cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane
PubChem CID 8728
IUPAC Name bromocyclopentane
SMILES C1CCC(C1)Br

Specifications

Color Yellow to Brown
Density 1.3900g/mL
Boiling Point 137°C to 139°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4881 to 1.4901
Linear Formula C5H9Br
Quantity 1 L
Beilstein 05, 19
Specific Gravity 1.39
Solubility Information Solubility in water: insoluble in water
Formula Weight 149.03
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclopentyl bromide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-294-6
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.