Learn More
Sigma Aldrich Fine Chemicals Biosciences Cyclopentyl methyl ether, 1L, ≥99.9%
Catalog No. NC1624695
Encompass
Cyclopentyl methyl ether Green Alternative contains 50 ppm BHT as inhibitor, anhydrous, ≥99.9% ** For R&D use only. Not for drug, household, or other uses.
This item is not returnable. View return policy
Catalog No. NC1624695 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 6759701L
Cyclopentyl methyl ether Green Alternative contains 50 ppm BHT as inhibitor, anhydrous, ≥99.9% ** For R&D use only. Not for drug, household, or other uses.