Learn More
Sigma Aldrich Fine Chemicals Biosciences Cyclopentyl methyl ether c
Catalog No. 502907857
Encompass
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. Cyclopentyl methyl ether (CPME) is a greener alternative to other ethereal solvents such as tetrahydrofuran (THF) 2-methyl tetrahydrofuran (2-MeTHF) dioxane (carcinogenic) tert-Butyl methyl ether (MTBE) and 12-dimethoxyethane (DME) and thus has been enhanced for Safer Solvents and Auxiliaries. Click here for more information.CPME is a greener alternative solvent for many aprotic ether solvents. It has many properties which make it safer and greener alternative .nbspHigh boiling point.nbspClean organic-water phase separation.nbspReduced energy to recoverIt also is safer for use with a lower peroxide formation than either THF or 2-MeTHF.
Catalog No. 50-290-7857 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 675989195L
Add to Cart
Add to Cart
Request a Quote
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. Cyclopentyl methyl ether (CPME) is a greener alternative to other ethereal solvents such as tetrahydrofuran (THF) 2-methyl tetrahydrofuran (2-MeTHF) dioxane (carcinogenic) tert-Butyl methyl ether (MTBE) and 12-dimethoxyethane (DME) and thus has been enhanced for Safer Solvents and Auxiliaries. Click here for more information.CPME is a greener alternative solvent for many aprotic ether solvents. It has many properties which make it safer and greener alternative .nbspHigh boiling point.nbspClean organic-water phase separation.nbspReduced energy to recoverIt also is safer for use with a lower peroxide formation than either THF or 2-MeTHF.