Learn More
Sigma Aldrich Fine Chemicals Biosciences Cyclopentyl methyl ether contains 50 ppm BHT as inhibitor, anhydrous, >=99.9% | 5614-37-9 | MFCD08276401 | 2L
Catalog No. 501746296
Encompass
Cyclopentyl methyl ether contains 50 ppm BHT as inhibitor, anhydrous, >=99.9% | Purity: >=99.9% | Mol Wt: 100.16 | 5614-37-9 | MFCD08276401 | 2L
This item is not returnable. View return policy
Catalog No. 50-174-6296 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 6759702L
Cyclopentyl methyl ether contains 50 ppm BHT as inhibitor, anhydrous, >=99.9% | Purity: >=99.9% | Mol Wt: 100.16 | 5614-37-9 | MFCD08276401 | 2L