Learn More
Medchemexpress LLC Cyclopentyl phenyl ketone | 5422-88-8 | MFCD00001378 | 98.4% | 174.24 g/mol | C12H14O | 10 G
SDP
Catalog No. 5000301211
Encompass_Preferred
Cyclopentyl phenyl ketone (CAS 5422-88-8) is an organic synthesis intermediate used as a building block in the preparation of active molecules and other fine chemicals. Provided for research use, it is characterized by a high purity and supports common ketone transformations such as reductions, condensations, and acylations.
  • High purity (98.42%).
  • Chemical formula C12H14O and molecular weight 174.24 g/mol.
  • Suitable as a building block for synthesis of active molecules and intermediates.
  • Supports common ketone transformations, e.g., reductions and condensations.
  • Supplied with certificate of analysis and safety data sheet for quality and safety assurance.
Catalog No. 50-003-01211 Supplier Medchemexpress LLC Supplier No. HYW01677910G
Cyclopentyl phenyl ketone (CAS 5422-88-8) is an organic synthesis intermediate used as a building block in the preparation of active molecules and other fine chemicals. Provided for research use, it is characterized by a high purity and supports common ketone transformations such as reductions, condensations, and acylations.
  • High purity (98.42%).
  • Chemical formula C12H14O and molecular weight 174.24 g/mol.
  • Suitable as a building block for synthesis of active molecules and intermediates.
  • Supports common ketone transformations, e.g., reductions and condensations.
  • Supplied with certificate of analysis and safety data sheet for quality and safety assurance.