Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclopentylboronic acid | 63076-51-7 | 113.95 | 25 G
SDP

Catalog No. 5000450088
Encompass_Preferred

Cyclopentylboronic acid is a biochemical reagent intended for life science related research, functioning as a biological material or organic compound. It is for research use only and not sold to patients.

  • Appearance: White to off-white solid
  • Molecular formula: C5H11BO2
  • Molecular weight: 113.95
  • Purity: ≥97.0% (NMR)
  • CAS number: 63076-51-7
  • Melting point: 126°C
  • Boiling point: 235°C at 760 mmHg

Catalog No. 50-004-50088 Supplier Medchemexpress LLC Supplier No. 25GHYW000955
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclopentylboronic acid is a biochemical reagent intended for life science related research, functioning as a biological material or organic compound. It is for research use only and not sold to patients.

  • Appearance: White to off-white solid
  • Molecular formula: C5H11BO2
  • Molecular weight: 113.95
  • Purity: ≥97.0% (NMR)
  • CAS number: 63076-51-7
  • Melting point: 126°C
  • Boiling point: 235°C at 760 mmHg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.