Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclophilin A antibody (YA481) | 18 kDa | 10 UL
SDP

Catalog No. 5000398974
Encompass_Preferred

Cyclophilin A Antibody (YA481) is a rabbit-derived, non-conjugated IgG monoclonal antibody, specifically designed to target Cyclophilin A. It is supplied as a liquid at a concentration of 1 mg/mL.

  • Host: Rabbit
  • Reactivity: Human, Mouse
  • Applications: WB, IHC-P
  • Formulation: 1*TBS (pH7.4), 0.05% BSA, 40% Glycerol, and 0.05% Sodium Azide
  • Storage: Store at -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Shipping: Shipped with blue ice

Catalog No. 50-003-98974 Supplier Medchemexpress LLC Supplier No. HYP8010110UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Cyclophilin A Antibody (YA481) is a rabbit-derived, non-conjugated IgG monoclonal antibody, specifically designed to target Cyclophilin A. It is supplied as a liquid at a concentration of 1 mg/mL.

  • Host: Rabbit
  • Reactivity: Human, Mouse
  • Applications: WB, IHC-P
  • Formulation: 1*TBS (pH7.4), 0.05% BSA, 40% Glycerol, and 0.05% Sodium Azide
  • Storage: Store at -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Shipping: Shipped with blue ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.