Learn More
Medchemexpress LLC Cyclophilin A antibody (YA481) | 18 kDa | 100 UL
SDP
Catalog No. 5000397519
Encompass_Preferred
Cyclophilin A antibody (YA481) is a rabbit-derived, non-conjugated IgG monoclonal antibody developed to target Cyclophilin A. It is supplied as a liquid at a concentration of 1 mg/mL.
  • Targets Cyclophilin A
  • Rabbit-derived IgG monoclonal antibody
  • Non-conjugated
  • Reacts with human and mouse samples
  • Applicable in Western Blot (WB) and Immunohistochemistry-Paraffin (IHC-P)
  • Formulated in 1*TBS (pH7.4), 0.05% BSA, 40% Glycerol, and 0.05% Sodium Azide
  • Predicted molecular weight of 18 kDa
  • Stable for 1 year when stored at -20°C (avoid freeze/thaw cycles)
  • Shipped with blue ice
Catalog No. 50-003-97519 Supplier Medchemexpress LLC Supplier No. HYP80101100UL
Cyclophilin A antibody (YA481) is a rabbit-derived, non-conjugated IgG monoclonal antibody developed to target Cyclophilin A. It is supplied as a liquid at a concentration of 1 mg/mL.
  • Targets Cyclophilin A
  • Rabbit-derived IgG monoclonal antibody
  • Non-conjugated
  • Reacts with human and mouse samples
  • Applicable in Western Blot (WB) and Immunohistochemistry-Paraffin (IHC-P)
  • Formulated in 1*TBS (pH7.4), 0.05% BSA, 40% Glycerol, and 0.05% Sodium Azide
  • Predicted molecular weight of 18 kDa
  • Stable for 1 year when stored at -20°C (avoid freeze/thaw cycles)
  • Shipped with blue ice