Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclophilin A protein, mouse | >95.0% | 100UG
SDP

Catalog No. 5000177408
Encompass_Preferred

Recombinant mouse Cyclophilin A (peptidyl-prolyl isomerase A) is supplied as a tag-free, sterile 0.22 μm-filtered solution in PBS with 10% glycerol for research use. The protein is expressed in E. coli, exhibits >95% purity by reducing SDS-PAGE, and has an approximate molecular weight of 17.0 kDa; the product is provided as a 100 μg vial and should be stored at -80°C to maintain stability.

  • Tag-free recombinant protein for native activity.
  • High purity (>95%) suitable for biochemical assays.
  • Supplied as a sterile, filtered solution in PBS with 10% glycerol.
  • Expressed in E. coli for efficient production.
  • Approximate molecular weight 17.0 kDa.
  • Store at -80°C and avoid repeated freeze-thaw cycles.
  • Intended for research use only.

Catalog No. 50-001-77408 Supplier Medchemexpress LLC Supplier No. HYP70007A100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse Cyclophilin A (peptidyl-prolyl isomerase A) is supplied as a tag-free, sterile 0.22 μm-filtered solution in PBS with 10% glycerol for research use. The protein is expressed in E. coli, exhibits >95% purity by reducing SDS-PAGE, and has an approximate molecular weight of 17.0 kDa; the product is provided as a 100 μg vial and should be stored at -80°C to maintain stability.

  • Tag-free recombinant protein for native activity.
  • High purity (>95%) suitable for biochemical assays.
  • Supplied as a sterile, filtered solution in PBS with 10% glycerol.
  • Expressed in E. coli for efficient production.
  • Approximate molecular weight 17.0 kDa.
  • Store at -80°C and avoid repeated freeze-thaw cycles.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.