Learn More
Medchemexpress LLC Cyclophilin A Mouse 10ug
SDP
Supplier:  Medchemexpress LLC HYP7421210UG
Encompass_Preferred
Cyclophilin A (PPIA) protein with integrin binding and peptidyl-prolyl cis-trans isomerase activity influences platelet aggregation and neuron differentiation It is found in the extracellular space and myelin sheath showing ubiquitous expression in various tissues including the liver and central nervous system PPIA is associated with cholangiocarcinoma and HIV disease suggesting its role in fundamental cellular processes Cyclophilin A Protein Mouse (His) is the recombinant mouse-derived Cyclophilin A protein expressed by E coli with C-His labeled tag
Catalog No. 50-001-78850