Learn More
Enzo Life Sciences Cyclophilin A polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA2960100
Encompass_Preferred
Immunogen: Recombinant human cyclophilin A conjugated to KLH. Source: Whole rabbit serum. Species reactivity: Human, Mouse, Rat. Applications: ICC, WB. Formulation: Liquid. Neat serum.
Catalog No. 50-201-1794