Learn More
Medchemexpress LLC Cyclophilin inhibitor 1 | 1509904-96-4 | 99.5% | 593.67 | 25 MG
SDP
Catalog No. 5000439659
Encompass_Preferred
Cyclophilin inhibitor 1 is a potent and orally bioavailable cyclophilin A inhibitor, with a Kd of 5 nM, and shows effective anti-HCV activity, with an EC50 of 98 nM for HCV 2a.
  • Potent and orally bioavailable cyclophilin A inhibitor.
  • Kd of 5 nM for cyclophilin A.
  • Effective anti-HCV activity, with an EC50 of 98 nM for HCV 2a.
  • Appearance: Solid, white to off-white.
Catalog No. 50-004-39659 Supplier Medchemexpress LLC Supplier No. 25MGHY112712
Cyclophilin inhibitor 1 is a potent and orally bioavailable cyclophilin A inhibitor, with a Kd of 5 nM, and shows effective anti-HCV activity, with an EC50 of 98 nM for HCV 2a.
  • Potent and orally bioavailable cyclophilin A inhibitor.
  • Kd of 5 nM for cyclophilin A.
  • Effective anti-HCV activity, with an EC50 of 98 nM for HCV 2a.
  • Appearance: Solid, white to off-white.