Learn More
Medchemexpress LLC Cyclophilin inhibitor 1 | 1509904-96-4 | 99.55% | 593.67 | 50 MG
SDP
Catalog No. 5000437474
Encompass_Preferred
Cyclophilin inhibitor 1 is a potent and orally bioavailable cyclophilin A inhibitor. It demonstrates effective anti-HCV activity.
  • Potent cyclophilin A inhibitor.
  • Orally bioavailable.
  • Effective anti-HCV activity (EC50 of 98 nM for HCV 2a).
Catalog No. 50-004-37474 Supplier Medchemexpress LLC Supplier No. HY11271250MG
Cyclophilin inhibitor 1 is a potent and orally bioavailable cyclophilin A inhibitor. It demonstrates effective anti-HCV activity.
  • Potent cyclophilin A inhibitor.
  • Orally bioavailable.
  • Effective anti-HCV activity (EC50 of 98 nM for HCV 2a).