Learn More
Medchemexpress LLC Cyclophilin inhibitor 1 | 1509904-96-4 | 99.6% | 593.67 | 1 MG
SDP
Catalog No. 5000437131
Encompass_Preferred
Cyclophilin inhibitor 1 is a potent and orally bioavailable cyclophilin A inhibitor, with a Kd of 5 nM, showing effective anti-HCV activity, with an EC50 of 98 nM for HCV 2a.
  • Potent cyclophilin A inhibitor with a Kd of 5 nM.
  • Orally bioavailable.
  • Shows effective anti-HCV activity with an EC50 of 98 nM for HCV 2a.
  • Solid, white to off-white.
Catalog No. 50-004-37131 Supplier Medchemexpress LLC Supplier No. HY1127121MG
Cyclophilin inhibitor 1 is a potent and orally bioavailable cyclophilin A inhibitor, with a Kd of 5 nM, showing effective anti-HCV activity, with an EC50 of 98 nM for HCV 2a.
  • Potent cyclophilin A inhibitor with a Kd of 5 nM.
  • Orally bioavailable.
  • Shows effective anti-HCV activity with an EC50 of 98 nM for HCV 2a.
  • Solid, white to off-white.