Learn More
Medchemexpress LLC Cyclophilin inhibitor 1 | 1509904-96-4 | 99.6% | 593.67 | 1 ML
SDP
Catalog No. 5000436917
Encompass_Preferred
Cyclophilin inhibitor 1 is a potent and orally bioavailable cyclophilin A inhibitor, demonstrating a Kd of 5 nM. It exhibits effective anti-HCV activity, with an EC50 of 98 nM for HCV 2a.
  • Potent cyclophilin A inhibitor.
  • Orally bioavailable.
  • Exhibits anti-HCV activity.
  • Kd of 5 nM.
  • EC50 of 98 nM for HCV 2a.
Catalog No. 50-004-36917 Supplier Medchemexpress LLC Supplier No. HY11271210MM/1ML
Cyclophilin inhibitor 1 is a potent and orally bioavailable cyclophilin A inhibitor, demonstrating a Kd of 5 nM. It exhibits effective anti-HCV activity, with an EC50 of 98 nM for HCV 2a.
  • Potent cyclophilin A inhibitor.
  • Orally bioavailable.
  • Exhibits anti-HCV activity.
  • Kd of 5 nM.
  • EC50 of 98 nM for HCV 2a.