Learn More
Medchemexpress LLC Cyclophilin inhibitor 1 | 1509904-96-4 | 99.6% | 593.67 | 10 MG
SDP
Catalog No. 5000437022
Encompass_Preferred
Cyclophilin inhibitor 1 is a potent and orally bioavailable cyclophilin A inhibitor, with a Kd of 5 nM, shows effective anti-HCV activity, with an EC50 of 98 nM for HCV 2a.
  • Potent and orally bioavailable cyclophilin A inhibitor.
  • Shows effective anti-HCV activity.
Catalog No. 50-004-37022 Supplier Medchemexpress LLC Supplier No. HY11271210MG
Add to Cart
Add to Cart
Cyclophilin inhibitor 1 is a potent and orally bioavailable cyclophilin A inhibitor, with a Kd of 5 nM, shows effective anti-HCV activity, with an EC50 of 98 nM for HCV 2a.
  • Potent and orally bioavailable cyclophilin A inhibitor.
  • Shows effective anti-HCV activity.