Learn More
Medchemexpress LLC Cyclophilin inhibitor 1 | 1509904-96-4 | 99.6% | 593.67 g/mol | C31H39N5O7 | 5 MG
SDP
Catalog No. 5000262458
Encompass_Preferred
Cyclophilin inhibitor 1 is a potent, orally bioavailable small-molecule inhibitor of Cyclophilin A. It binds Cyclophilin A with a reported Kd of 5 nM and shows antiviral activity against hepatitis C virus genotype 2a with an EC50 of 98 nM. The compound is supplied as a solid with high purity and is soluble in DMSO for use in biochemical and cellular studies.
  • Potent Cyclophilin A inhibitor (Kd ~5 nM).
  • Demonstrated anti-HCV activity (EC50 98 nM, HCV 2a).
  • Reported oral bioavailability in preclinical studies.
  • High purity suitable for biochemical and cellular assays.
  • Soluble in DMSO; supplied as a solid for reconstitution.
  • Offered in small quantities for research use.
Catalog No. 50-002-62458 Supplier Medchemexpress LLC Supplier No. HY1127125MG
Cyclophilin inhibitor 1 is a potent, orally bioavailable small-molecule inhibitor of Cyclophilin A. It binds Cyclophilin A with a reported Kd of 5 nM and shows antiviral activity against hepatitis C virus genotype 2a with an EC50 of 98 nM. The compound is supplied as a solid with high purity and is soluble in DMSO for use in biochemical and cellular studies.
  • Potent Cyclophilin A inhibitor (Kd ~5 nM).
  • Demonstrated anti-HCV activity (EC50 98 nM, HCV 2a).
  • Reported oral bioavailability in preclinical studies.
  • High purity suitable for biochemical and cellular assays.
  • Soluble in DMSO; supplied as a solid for reconstitution.
  • Offered in small quantities for research use.