Learn More
TARGETMOL CHEMICALS INC CYCLOPHOSPHAMIDE 500MG
SDP
Catalog No. 502581781
Encompass_Preferred
Also available in 50 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. Cyclophosphamide is an alkylating agent used in the treatment of several forms of cancer including leukemias lymphomas and breast cancer. purity: 98%
Catalog No. 50-258-1781 Supplier TARGETMOL CHEMICALS INC Supplier No. T0707L500MG
Also available in 50 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. Cyclophosphamide is an alkylating agent used in the treatment of several forms of cancer including leukemias lymphomas and breast cancer. purity: 98%