Learn More
Cayman Chemical Cyclophosphamide-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 173547-45-0, 1 mg
SDP
Catalog No. 502987283
Encompass_Preferred
An internal standard for the quantification of cyclophosphamide by GC- or LC-MS
Catalog No. 50-298-7283 Supplier Cayman Chemical Supplier No. 221141 MG
An internal standard for the quantification of cyclophosphamide by GC- or LC-MS