Learn More
Apexbio Technology LLC Cyclophosphamide monohydrate 6055-19-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062657
Encompass_Preferred
Cyclophosphamide monohydrate is an alkylating cytotoxic agent exhibiting antitumor activity primarily via metabolic activation As a prodrug it undergoes hepatic biotransformation mediated by cytochrome P450 enzymes forming active alkylating metabolites such as 4-hydroxycyclophosphamide which exert cytotoxic effects through interactions with cellular DNA Cyclophosphamide also reversibly inhibits acetylcholinesterase (AChE) displaying an IC50 of approximately 511 M It is widely utilized in experimental cancer studies particularly in pediatric oncology where alterations in drug metabolism or drug-drug interactions impacting cyclophosphamide clearance and therapeutic outcomes have been explored
Catalog No. 50-000-62657 Supplier Apexbio Technology LLC Supplier No. A42325.1
Cyclophosphamide monohydrate is an alkylating cytotoxic agent exhibiting antitumor activity primarily via metabolic activation As a prodrug it undergoes hepatic biotransformation mediated by cytochrome P450 enzymes forming active alkylating metabolites such as 4-hydroxycyclophosphamide which exert cytotoxic effects through interactions with cellular DNA Cyclophosphamide also reversibly inhibits acetylcholinesterase (AChE) displaying an IC50 of approximately 511 M It is widely utilized in experimental cancer studies particularly in pediatric oncology where alterations in drug metabolism or drug-drug interactions impacting cyclophosphamide clearance and therapeutic outcomes have been explored