Learn More
Apexbio Technology LLC Cyclophosphamide monohydrate, 50mg. Cas: 6055-19-2 MFCD: MFCD00149395. alkylating, cytotoxic agent,antitumor activity
SDP
Supplier:  Apexbio Technology LLC A423250
Encompass_Preferred
IC50: 511 M for AChECyclophosphamide monohydrate is used in the cancer treatment in children. As a prodrug it should be metabolized by cytochrome P-450 enzymes to produce the active alkylating species, which are responsible for its cytotoxic effects. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4909