Learn More
Medchemexpress LLC HY-P7309 10ug Medchemexpress, TWEAK (CHO-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 501970595
Encompass_Preferred
Medchemexpress, HY-17420A 100mg Cyclophosphamide (hydrate) CAS:6055-19-2 Cyclophosphamide hydrate is a synthetic alkylating agent chemically related to the nitrogen mustards with antineoplastic and immunosuppressive activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0595 Supplier Medchemexpress LLC Supplier No. HYP730910UG
Medchemexpress, HY-17420A 100mg Cyclophosphamide (hydrate) CAS:6055-19-2 Cyclophosphamide hydrate is a synthetic alkylating agent chemically related to the nitrogen mustards with antineoplastic and immunosuppressive activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.