Learn More
Medchemexpress LLC Cyclophosphamide hydrate | 6055-19-2 | 99.9% | 500 MG
SDP
Supplier:  Medchemexpress LLC HY17420A500MG
Encompass_Preferred
Cyclophosphamide hydrate is a synthetic alkylating agent chemically related to the nitrogen mustards with antineoplastic and immunosuppressive activities. It induces outer membrane blebbing, leads to DNA fragmentation, and induces cleavage of caspase 3 and caspase 7 substrate PARP in 9L/P450 cells. In C3H mice bearing SW1 tumors, it increased the percentage of cells that stained for CD3, CD4, or CD8 in both spleens and tumors.
  • Acts as a synthetic alkylating agent
  • Exhibits antineoplastic and immunosuppressive activities
  • Induces DNA fragmentation and caspase cleavage
  • Increases immune cell markers in spleens and tumors
Catalog No. 50-003-85649