Learn More
Selleck Chemical LLC Cyclophosphamide Monohydrate 50mg 6055-19-2
SDP
Supplier:  Selleck Chemical LLC S205750MG
Encompass_Preferred
Cyclophosphamide Monohydrate is a nitrogen mustard alkylating agent, it attaches the alkyl group to the guanine base of DNA, shown to crosslink DNA, causing strand breakage and inducing mutations. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1222105