Learn More
Selleck Chemical LLC Cyclophosphamide Monohydrate S2057-200mg
SDP
Catalog No. 503108417
Encompass_Preferred
Cyclophosphamide Monohydrate is a nitrogen mustard alkylating agent it attaches the alkyl group to the guanine base of DNA shown to crosslink DNA causing strand breakage and inducing mutations Cyclophosphamide (NSC-26271) Monohydrate can be used to induce animal models of Anemia
Catalog No. 50-310-8417 Supplier Selleck Chemical LLC Supplier No. S2057200MG
Cyclophosphamide Monohydrate is a nitrogen mustard alkylating agent it attaches the alkyl group to the guanine base of DNA shown to crosslink DNA causing strand breakage and inducing mutations Cyclophosphamide (NSC-26271) Monohydrate can be used to induce animal models of Anemia