Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclophosphamide monohydrate, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111019890 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9890 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9890 Supplier MilliporeSigma Supelco Supplier No. 93813100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 6055-19-2
Flash Point 113°C
Physical Form Neat
Linear Formula C7H15 Cl2N2O2 P · H2O
MDL Number MFCD00149395
Melting Point 47°C to 52°C; 49°C to 51°C
Synonym 2-[Bis(2-chloroethyl)amino]tetrahydro-2 H-1,3,2-oxazaphosphorine 2-oxide; Cytoxan
Type Analytical Standard
Quantity 100 mg
Formula Weight 279.10
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.