Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclophosphamide Monohydrate, MP Biomedicals

Catalog No. ICN15074910 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15074910 10 g
ICN15074905 5 g
ICN15074925 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15074910 Supplier MP Biomedicals, Inc Supplier No. 0215074910
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6055-19-2
Molecular Formula C7H17Cl2N2O3P
Molecular Weight (g/mol) 279.10
MDL Number MFCD00149395
InChI Key PWOQRKCAHTVFLB-UHFFFAOYNA-N
Synonym cyclophosphamide monohydrate, cyclophosphamide hydrate, cytoxan, endoxon, endoxan monohydrate, ciclophosphamide hydrat, cytoxan hydrate, endoxan a, cyclophosphamide hydrated
PubChem CID 22420
ChEBI CHEBI:4026
IUPAC Name 2-[bis(2-chloroethyl)amino]-1,3,2λ⁵-oxazaphosphinan-2-one hydrate
SMILES O.ClCCN(CCCl)P1(=O)NCCCO1

Specifications

Assay Percent Range >98%
Quantity 10 g
Formula Weight 279.1
Chemical Name or Material Cyclophosphamide Monohydrate
Recommended Storage Store at + 4°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.