Learn More
Cayman Chemical CyclopIazonIc AcId 10mg
SDP
Catalog No. 502951605
Encompass_Preferred
A mycotoxin and SERCA inhibitor (IC50 = 0.6 µM); induces ptosis, hypothermia, tremor, and cachexia, as well as induces lethality (LD50 = 13 mg/kg), in rats from 11-14 mg/kg
Catalog No. 50-295-1605 Supplier Cayman Chemical Supplier No. 1132610 MG
A mycotoxin and SERCA inhibitor (IC50 = 0.6 µM); induces ptosis, hypothermia, tremor, and cachexia, as well as induces lethality (LD50 = 13 mg/kg), in rats from 11-14 mg/kg