Learn More
Cayman Chemical Cyclopiazonic Acid-13C20, Purity: ≥98%, 1.2 mL
SDP
Catalog No. 502990012
Encompass_Preferred
An internal standard for the quantification of cyclopiazonic acid by GC- or LC-MS
Catalog No. 50-299-0012 Supplier Cayman Chemical Supplier No. 312971.2 ML
An internal standard for the quantification of cyclopiazonic acid by GC- or LC-MS