Learn More
Medchemexpress LLC Cyclopiazonic acid | 18172-33-3 | 10 MG
SDP
Catalog No. 5000419306
Encompass_Preferred
Cyclopiazonic acid is a potent endoplasmic reticulum calcium ATPase (ECAs) inhibitor and a human respiratory syncytial virus (RSV) inhibitor, with an EC50 value of 4.13 μM. It plays a role in reducing the antagonistic effect of 5-HT receptors in rat thoracic aorta, inducing p53 dependent cell apoptosis and reproductive toxicity in mouse testes, and inhibiting the biological activation of aflatoxin B.
  • Functions as an ECAs inhibitor
  • Acts as an RSV inhibitor (EC50 value of 4.13 μM)
  • Reduces antagonistic effects of 5-HT receptors
  • Induces p53 dependent cell apoptosis
  • Inhibits biological activation of aflatoxin B
  • Off-white to yellow solid appearance
  • High purity of 99.86%
  • Soluble in DMSO at 100 mg/mL
Catalog No. 50-004-19306 Supplier Medchemexpress LLC Supplier No. HYN677110MG
Cyclopiazonic acid is a potent endoplasmic reticulum calcium ATPase (ECAs) inhibitor and a human respiratory syncytial virus (RSV) inhibitor, with an EC50 value of 4.13 μM. It plays a role in reducing the antagonistic effect of 5-HT receptors in rat thoracic aorta, inducing p53 dependent cell apoptosis and reproductive toxicity in mouse testes, and inhibiting the biological activation of aflatoxin B.
  • Functions as an ECAs inhibitor
  • Acts as an RSV inhibitor (EC50 value of 4.13 μM)
  • Reduces antagonistic effects of 5-HT receptors
  • Induces p53 dependent cell apoptosis
  • Inhibits biological activation of aflatoxin B
  • Off-white to yellow solid appearance
  • High purity of 99.86%
  • Soluble in DMSO at 100 mg/mL