Learn More
Enzo Life Sciences Cyclopiazonic acid (50mg). CAS: 18172-33-3
SDP
Supplier:  Enzo Life Sciences BMLCA4150050
Encompass_Preferred
Mycotoxin. Cell permeable. Induces the release of intracellular stored Ca2+ without increasing IP3 levels via inhibition of endoplasmic reticulum Ca2+-ATPase. Highly specific inhibitor of Ca2+-ATPase of sarcoplasmic reticulum, completely inhibiting the enzyme at 6-8 nmol/mg protein (at 0.5-2.0µM ATP). Alternative name: CPA. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (20mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0394