Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopiazonic Acid, >98%, MP Biomedicals

Catalog No. ICN15889550 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15889550 50 mg
ICN15889505 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15889550 Supplier MP Biomedicals, Inc Supplier No. 0215889550
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18172-33-3
Molecular Formula C20H20N2O3
Molecular Weight (g/mol) 336.391
InChI Key SZINUGQCTHLQAZ-AFAVFJNCSA-N
Synonym cyclopiazonic acid, alpha-cyclopiazonic acid, unii-x9tly4580z, ccris 4942, alpha-cyclopiazonate, cyclopyazonic acid, 6ar,10z,11as,11br-10-1-hydroxyethylidene-7,7-dimethyl-6a,7,11a,11b-tetrahydro-6h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indole-9,11 2h,10h-dione, 9h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-9-one, 2,6,6a,7,11a,11b-hexahydro-10-acetyl-7,7-dimethyl-11-hydroxy-, 6a-alpha,11a,11b-alpha, 6ar,11as,11br-10-acetyl-9-hydroxy-7,7-dimethyl-2,6,6a,7,11a,11b-hexahydro-11h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-11-one, ambotzls-1020
PubChem CID 54711281
SMILES CC(=O)C1=C(C2C3C(CC4=C5C3=CNC5=CC=C4)C(N2C1=O)(C)C)O

Specifications

Color White
Assay Percent Range >98%
Quantity 50 mg
Formula Weight 336.4
Physical Form Powder
Chemical Name or Material Cyclopiazonic Acid
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.