Learn More
Abcam Cyclopiazonic acid from Penicillium grisefulvum, Ca2+-ATPase inhibitor, 50MG
Catalog No. 502707703
Encompass_Preferred
MW 336.4 Da, Purity >99%. Cell-permeable, reversible inhibitor of sarcoplasmic reticulum Ca2+-ATPase.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7703 Supplier Abcam Supplier No. AB12030050MG
MW 336.4 Da, Purity >99%. Cell-permeable, reversible inhibitor of sarcoplasmic reticulum Ca2+-ATPase.
The product is subject to the following: Abcam Restricted Use Statement