Learn More
Abcam Cyclopiazonic acid from Penicillium grisefulvum, Ca2+-ATPase inhibitor, 10MG
Catalog No. 502707704
Encompass_Preferred
MW 336.4 Da, Purity >99%. Cell-permeable, reversible inhibitor of sarcoplasmic reticulum Ca2+-ATPase.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7704 Supplier Abcam Supplier No. AB12030010MG
MW 336.4 Da, Purity >99%. Cell-permeable, reversible inhibitor of sarcoplasmic reticulum Ca2+-ATPase.
The product is subject to the following: Abcam Restricted Use Statement