Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopiazonic Acid, MP Biomedicals™

Catalog No. MP215889510 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215889510 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215889510 Supplier MP Biomedicals, Inc Supplier No. 0215889510
Add to Cart
Add to Cart

Displays anti-arrhythmic effects. Cyclopiazonic Acid, MP Biomedicals inhibits the sarcoplasmic reticulum Ca2+-ATPase, which induces the release of intracellular stored Ca2+ without increasing IP3 levels.

Cyclopiazonic acid is a highly specific inhibitor of the Ca2+-ATPase of sarcoplasmic reticulum, completely inhibiting the enzyme at 6-8nmol/mg protein at the low concentration of 0.5-2μM. Cyclopiazonic acid on Langendorff-perfused murine hearts displays anti-arrhythmic effects. Also, administration on rat soleus muscle showed a decrease in hormone-sensitive lipase activity and triacylglycerol hydrolysis.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18172-33-3
Molecular Formula C20H20N2O3
Molecular Weight (g/mol) 336.391
InChI Key SZINUGQCTHLQAZ-AFAVFJNCSA-N
Synonym cyclopiazonic acid, alpha-cyclopiazonic acid, unii-x9tly4580z, ccris 4942, alpha-cyclopiazonate, cyclopyazonic acid, 6ar,10z,11as,11br-10-1-hydroxyethylidene-7,7-dimethyl-6a,7,11a,11b-tetrahydro-6h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indole-9,11 2h,10h-dione, 9h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-9-one, 2,6,6a,7,11a,11b-hexahydro-10-acetyl-7,7-dimethyl-11-hydroxy-, 6a-alpha,11a,11b-alpha, 6ar,11as,11br-10-acetyl-9-hydroxy-7,7-dimethyl-2,6,6a,7,11a,11b-hexahydro-11h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-11-one, ambotzls-1020
PubChem CID 54711281
SMILES CC(=O)C1=C(C2C3C(CC4=C5C3=CNC5=CC=C4)C(N2C1=O)(C)C)O

Specifications

Melting Point 237°C to 238°C (literature)
Quantity 10 mg
Solubility Information Soluble in chloroform (10mg/mL) or methanol (10mg/mL, clear, bright yellow solution).
Formula Weight 336.4
Percent Purity >98%
Physical Form Powder
Recommended Storage −20°C. Store Desiccated.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.