Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclopropanecarbonyl chloride

Catalog No. C116807100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C116807100G 100 g
C11680725G 25 g
C1168075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C116807100G Supplier Sigma Aldrich Supplier No. C116807100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cyclopropanecarbonyl chloride can be used as an alkylating reagent in the preparation of some chemical building blocks such as formylcyclopropane, cyclopropanecarboxylic acid hydrazide, bis(cyclopropylcarbonyl) peroxide, 1-cyclopropanecarboxamide, 2-cyclopropylcarbonylcyclohexane-1,3-diones and cyclopropyl analog of triazolopiperazine-amides.

Specifications

CAS 4023-34-1
Density 1.152 g/mL (at 25°C (literature))
Boiling Point 119°C (lit.)
Molecular Formula C4H5ClO
Linear Formula C3H5COCl
Refractive Index n20/D 1.452 (literature)
MDL Number MFCD00001277
Quantity 100 g
Synonym Cyclopropanecarboxylic acid chloride
Molecular Weight (g/mol) 104.53
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.