Learn More
Medchemexpress LLC Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propen-1-yl)-, (3-phenoxyphenyl)methyl... | 26002-80-2 | 94.6% | 25 MG
SDP
Catalog No. 5000414515
Encompass_Preferred
Phenothrin is a Type I pyrethroid insecticide that induces dose-dependent DNA damage in human peripheral blood lymphocytes and hepatocytes, and exhibits definite genotoxic potential. It is used for pest control in agriculture, households, public health, and for eliminating human head lice infestations.
  • Type I pyrethroid insecticide
  • Induces dose-dependent DNA damage
  • Used for pest control in agriculture, households, public health
  • Eliminates human head lice infestations
  • Available in pure form for research use
Catalog No. 50-004-14515 Supplier Medchemexpress LLC Supplier No. 25MGHYB1072
Phenothrin is a Type I pyrethroid insecticide that induces dose-dependent DNA damage in human peripheral blood lymphocytes and hepatocytes, and exhibits definite genotoxic potential. It is used for pest control in agriculture, households, public health, and for eliminating human head lice infestations.
  • Type I pyrethroid insecticide
  • Induces dose-dependent DNA damage
  • Used for pest control in agriculture, households, public health
  • Eliminates human head lice infestations
  • Available in pure form for research use