Learn More
Medchemexpress LLC Cyclopropanecarboxylic acid, 3-(2,2-dibromoethenyl)-2,2-dimethyl-, (S)-cyano (3-phenoxyphenyl)methyl ester, (1R,3R)- | 52918-63-5 | 99.9% | 100 MG
SDP
Catalog No. 5000410088
Encompass_Preferred
Deltamethrin is an orally active synthetic pyrethroid insecticide that induces oxidative stress and results in inflammation and apoptosis by inhibiting the Nrf2/HO-1 pathway. It also has an anticancer effect by inducing apoptosis and is used extensively in pest control.
  • Orally active synthetic pyrethroid insecticide
  • Induces oxidative stress
  • Results in inflammation and apoptosis via inhibiting Nrf2/HO-1 pathway
  • Has an anticancer effect by inducing apoptosis
  • Can be used extensively in pest control
Catalog No. 50-004-10088 Supplier Medchemexpress LLC Supplier No. HYB1971100MG
Deltamethrin is an orally active synthetic pyrethroid insecticide that induces oxidative stress and results in inflammation and apoptosis by inhibiting the Nrf2/HO-1 pathway. It also has an anticancer effect by inducing apoptosis and is used extensively in pest control.
  • Orally active synthetic pyrethroid insecticide
  • Induces oxidative stress
  • Results in inflammation and apoptosis via inhibiting Nrf2/HO-1 pathway
  • Has an anticancer effect by inducing apoptosis
  • Can be used extensively in pest control