Learn More
Medchemexpress LLC Cyclopropanecarboxylic acid, 3-(2,2-dibromoethenyl)-2,2-dimethyl-, (S)-cyano (3-phenoxyphenyl)methyl ester | 52918-63-5 | 99.9% | 500 MG
SDP
Catalog No. 5000410528
Encompass_Preferred
Deltamethrin (Decamethrin) is an orally active synthetic pyrethroid insecticide. It induces oxidative stress, inflammation, and apoptosis by inhibiting the Nrf2/HO-1 pathway. Deltamethrin also has an anticancer effect by inducing apoptosis and is extensively used in pest control.
  • Orally active synthetic pyrethroid insecticide.
  • Induces oxidative stress.
  • Results in inflammation and apoptosis via inhibiting Nrf2/HO-1 pathway.
  • Has an anticancer effect by inducing apoptosis.
  • Can be used extensively in pest control.
Catalog No. 50-004-10528 Supplier Medchemexpress LLC Supplier No. HYB1971500MG
Deltamethrin (Decamethrin) is an orally active synthetic pyrethroid insecticide. It induces oxidative stress, inflammation, and apoptosis by inhibiting the Nrf2/HO-1 pathway. Deltamethrin also has an anticancer effect by inducing apoptosis and is extensively used in pest control.
  • Orally active synthetic pyrethroid insecticide.
  • Induces oxidative stress.
  • Results in inflammation and apoptosis via inhibiting Nrf2/HO-1 pathway.
  • Has an anticancer effect by inducing apoptosis.
  • Can be used extensively in pest control.