Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclopropanemethanol

Catalog No. 48209925ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-209-925ML 25 mL
48-209-91L 1 L
48-209-9250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 48-209-925ML Supplier Sigma Aldrich Supplier No. 48209925ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Cyclopropanemethanol (cyclopropyl carbinol, cpmo), a cycloalkanemethanol, is an anaesthetic. the coupling reaction of cyclopropanemethanol with alkynes to form substituted allylic alcohols has been reported. the microwave spectrum of cpmo has been recorded. its rotational constants and dipole moment have been determined.cyclopropanemethanol may be used in the preparation of:
  • Cyclopropanecarbaldehyde
  • Cyclopropylmethylsulfonate
  • Dibenzyl cyclopropylmethyl phosphate

Specifications

CAS 2516-33-8
Density 0.89 g/mL (at 25°C (literature))
Boiling Point 123°C to 124°C (738 mmHg)
Molecular Formula C4H8O
Refractive Index n20/D 1.431 (literature)
Linear Formula C3H5CH2OH
MDL Number MFCD00001309
Quantity 25 mL
Synonym (Hydroxymethyl)cyclopropane; CPMO; Cyclopropyl carbinol; Cyclopropylmethanol
Molecular Weight (g/mol) 72.11
Percent Purity ≥99.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.