Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropanesulfonamide 98.0+%, TCI America™
SDP

Catalog No. C26285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26285G 5 g
C26281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26285G Supplier TCI America Supplier No. C26285G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 154350-29-5
Molecular Formula C3H7NO2S
Molecular Weight (g/mol) 121.15
MDL Number MFCD08705286
InChI Key WMSPXQIQBQAWLL-UHFFFAOYSA-N
PubChem CID 15765418
IUPAC Name cyclopropanesulfonamide
SMILES NS(=O)(=O)C1CC1

Specifications

Melting Point 108°C
Color White
Quantity 5 g
Formula Weight 121.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cyclopropanesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.