Learn More
Medchemexpress LLC Cyclopropanesulfonyl chloride | 139631-62-2 | 140.59 | 10 G
SDP
Catalog No. 5000449138
Encompass_Preferred
Cyclopropanesulfonyl chloride is an intermediate that can be used to synthesize Compound 1. Compound 1 has anticancer activity against EGFR triple-mutant tumors.
  • Used for research purposes only.
  • Can be used to synthesize Compound 1, which has anticancer activity against EGFR triple-mutant tumors.
  • Purity of 99.9%.
Catalog No. 50-004-49138 Supplier Medchemexpress LLC Supplier No. HYW00279810G
Cyclopropanesulfonyl chloride is an intermediate that can be used to synthesize Compound 1. Compound 1 has anticancer activity against EGFR triple-mutant tumors.
  • Used for research purposes only.
  • Can be used to synthesize Compound 1, which has anticancer activity against EGFR triple-mutant tumors.
  • Purity of 99.9%.