Learn More
Medchemexpress LLC Cyclopropavir | 632325-71-4 | 98.0% | 263.26 | 50 MG
SDP
Catalog No. 5000393892
Encompass_Preferred
Cyclopropavir (Filociclovir) is an orally active and broad-spectrum anti-herpesvirus compound. It inhibits UL97 kinase activity and has antiviral activity against HCMV, MCMV, and HAdV6 viruses.
  • Orally active compound
  • Broad-spectrum anti-herpesvirus activity
  • Inhibits UL97 kinase activity
  • Active against HCMV, MCMV, and HAdV6 viruses
Catalog No. 50-003-93892 Supplier Medchemexpress LLC Supplier No. HY1672150MG
Cyclopropavir (Filociclovir) is an orally active and broad-spectrum anti-herpesvirus compound. It inhibits UL97 kinase activity and has antiviral activity against HCMV, MCMV, and HAdV6 viruses.
  • Orally active compound
  • Broad-spectrum anti-herpesvirus activity
  • Inhibits UL97 kinase activity
  • Active against HCMV, MCMV, and HAdV6 viruses