Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropyl 2-Thienyl Ketone 96.0+%, TCI America™
SDP

Catalog No. C108810G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C108810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C108810G Supplier TCI America Supplier No. C108810G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6193-47-1
Molecular Formula C8H8OS
Molecular Weight (g/mol) 152.211
MDL Number MFCD00005427
InChI Key PJDFNFSTSCAKPC-UHFFFAOYSA-N
Synonym cyclopropyl 2-thienyl ketone, cyclopropyl thiophen-2-yl methanone, cyclopropyl 2-thienyl methanone, cyclopropyl2-thienylketone, acmc-1b3oh, 2-thienylcyclopropyl ketone, cyclopropyl 2-thienylketone, bidd:gt0674, methanone,cyclopropyl-2-thienyl, pjdfnfstscakpc-uhfffaoysa
PubChem CID 80328
IUPAC Name cyclopropyl(thiophen-2-yl)methanone
SMILES C1CC1C(=O)C2=CC=CS2

Specifications

Color Yellow
Boiling Point 120°C
Quantity 10 g
Formula Weight 152.21
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopropyl 2-Thienyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.