Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropyl 4-methoxyphenyl Ketone 98%, Thermo Scientific™

Catalog No. AC111690250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC111690250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC111690250 Supplier Thermo Scientific Chemicals Supplier No. 111690250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7152-03-6
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.22
InChI Key YKZSVEVTRUSPOQ-UHFFFAOYSA-N
Synonym cyclopropyl 4-methoxyphenyl methanone, cyclopropyl 4-methoxyphenyl ketone, methanone, cyclopropyl 4-methoxyphenyl, chembl82673, cyclopropyl-4-methoxyphenyl methanone, cyclopropyl-4-methoxy-phenyl-methanone, maybridge3_006117, 4-methoxybenzoylcyclopropane, cyclopropyl p-anisyl ketone, methanone,cyclopropyl 4-methoxyphenyl
PubChem CID 81580
IUPAC Name cyclopropyl-(4-methoxyphenyl)methanone
SMILES COC1=CC=C(C=C1)C(=O)C2CC2

Specifications

Melting Point 38°C to 44°C
Color Brown to Yellow
Infrared Spectrum Authentic
Linear Formula CH3OC6H4COC3H5
Quantity 25 g
Formula Weight 176.21
Percent Purity ≥97.5%
Physical Form Crystalline Solid
Chemical Name or Material Cyclopropyl 4-methoxyphenyl ketone, 98%
Hazard Category Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.