Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropyl Carbinol, 99%

Catalog No. ac296820250
Change view
Click to view available options
Quantity:
25 mL
100 mL
250 mL
1 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296820250 25 mL Glass bottle
AC296821000 100 mL Glass bottle
AC296822500 250 mL Glass bottle
AC296820010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC296820250 Supplier Thermo Scientific Chemicals Supplier No. 296820250
Only null left
Add to Cart

CAS: 2516-33-8 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2516-33-8
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00001309
InChI Key GUDMZGLFZNLYEY-UHFFFAOYSA-N
Synonym cyclopropanemethanol, cyclopropyl carbinol, cyclopropylcarbinol, cyclopropylmethyl alcohol, hydroxymethyl cyclopropane, cyclopropanemethyl alcohol, cyclopropylcarbinyl alcohol, hydroxymethylcyclopropane, cyclopropyl-methanol, cyclopropane methanol
PubChem CID 75644
IUPAC Name cyclopropylmethanol
SMILES OCC1CC1

Specifications

Melting Point -60°C
Color Colorless
Density 0.8900g/mL
Boiling Point 123°C to 124°C (738.0 mmHg)
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4310 to 1.433
Linear Formula C3H5CH2OH
Quantity 25 mL
Beilstein 06,4
Specific Gravity 0.89
Solubility Information Solubility in water: miscible. Other solubilities: miscible with ethanol,slightly soluble in carbon tetrachloride
Viscosity 4.04 mPa.s (20°C)
Formula Weight 72.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cyclopropyl carbinol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-735-5
RTECSNumber GZ2212250
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.