Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropyl Diphenyl Carbinol, 99+%, Thermo Scientific™

Catalog No. AC111670050
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC111670050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC111670050 Supplier Thermo Scientific Chemicals Supplier No. 111670050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5785-66-0
Infrared Spectrum Authentic
Assay Percent Range 99+%
Molecular Formula C16H16O
Linear Formula C3H5C(C6H5)2OH
Quantity 5g
MDL Number MFCD00001298
Synonym cyclopropyldiphenylcarbinol, cyclopropyl diphenyl carbinol, methanol, cyclopropyldiphenyl, alpha-cyclopropylbenzhydrylic alcohol, cyclopropyl diphenyl methanol, diphenyl cyclopropyl carbinol, alpha-cyclopropyl-alpha-phenylbenzyl alcohol, benzyl alcohol, alpha-cyclopropyl-alpha-phenyl, benzenemethanol, alpha-cyclopropyl-alpha-phenyl-9ci, benzenemethanol, alpha-cyclopropyl-alpha-phenyl
InChI Key MFKPHBJFWOOEDT-UHFFFAOYSA-N
SMILES OC(C1CC1)(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name cyclopropyldiphenylmethanol
Molecular Weight (g/mol) 224.30
PubChem CID 79850
Formula Weight 224.3
Percent Purity ≥99.0%
Chemical Name or Material Cyclopropyl diphenyl carbinol, 99+%
Show More Show Less
EINECSNumber 227-312-1
RTECSNumber PC1950000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.