Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclopropyl methyl ketone

Catalog No. C12000625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12000625G 25 g
C120006100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12000625G Supplier Sigma Aldrich Supplier No. C12000625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cyclopropyl methyl ketone

Specifications

CAS 765-43-5
Density 0.849 g/mL (at 25°C (literature))
Boiling Point 114°C (lit.)
Molecular Formula C5H8O
Refractive Index n20/D 1.424 (literature)
Linear Formula C3H5COCH3
MDL Number MFCD00001297
Quantity 25 g
Synonym Acetylcyclopropane
Molecular Weight (g/mol) 84.12
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.