Learn More
Medchemexpress LLC Cyclopropyl (S)-2-(4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f]triazolo[4,3-a]diazepin-6-yl) | 3077900-08-1 | 98.3% | 440.95 | C22H21ClN4O2S | 10 MG
SDP
Catalog No. 5000294895
Encompass_Preferred
DDL-357 is a small-molecule secreted clusterin (sCLU) enhancer used as a research probe to increase sCLU levels in brain, reduce pathological phospho-tau, and improve memory in Alzheimer's disease mouse models. It is supplied as a 10 mg research sample intended for preclinical and in vitro studies.
  • Potent secreted clusterin enhancer that increases sCLU in brain.
  • Reduces phospho-tau and improves cognitive performance in murine 3xTg-AD models.
  • Suitable for preclinical in vivo and in vitro research applications.
  • Provided as a 10 mg research sample with 98.3% purity.
  • Molecular weight 440.95; molecular formula C22H21ClN4O2S; CAS 3077900-08-1.
Catalog No. 50-002-94895 Supplier Medchemexpress LLC Supplier No. HY17643810MG
DDL-357 is a small-molecule secreted clusterin (sCLU) enhancer used as a research probe to increase sCLU levels in brain, reduce pathological phospho-tau, and improve memory in Alzheimer's disease mouse models. It is supplied as a 10 mg research sample intended for preclinical and in vitro studies.
  • Potent secreted clusterin enhancer that increases sCLU in brain.
  • Reduces phospho-tau and improves cognitive performance in murine 3xTg-AD models.
  • Suitable for preclinical in vivo and in vitro research applications.
  • Provided as a 10 mg research sample with 98.3% purity.
  • Molecular weight 440.95; molecular formula C22H21ClN4O2S; CAS 3077900-08-1.