Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclopropylamine

Catalog No. 12550425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-550-425G 25 g
12-550-410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-550-425G Supplier Sigma Aldrich Supplier No. 12550425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Cyclopropylamine(cpa) has been used in the synthesis of n-[4-(4-fluoro)phenyl-2-aminothiazol-5-yl]pyrimidin-2-yl-alkylamine derivatives
  • It has been used in the synthesis of pt(cpa)2(bismethylthiomethylenepropanedioate) and pt(cpa)2(bisethylthiomethylenepropanedioate) complexes.cyclopropylamine inactivates cytochrome p450 enzymes by a mechanism involving initial one-electron oxidation at nitrogen followed by scission of the cyclopropane ring leading to covalent modification of the enzyme
  • It is a mechanism-based inhibitor of qui

Specifications

CAS 765-30-0
Density 0.824 g/mL (at 25°C (literature))
Boiling Point 49°C to 50°C (lit.)
Molecular Formula C3H7N
Refractive Index n20/D 1.420 (literature)
Linear Formula C3H5NH2
MDL Number MFCD00001301
Quantity 25 g
Synonym Aminocyclopropane
Molecular Weight (g/mol) 57.09
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.